Product Details

SNP ID
rs37360
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:58164911 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGACTTTAAGGGGTGAACGTTTCC[A/C]TCTTGATTCAGAAAGCGACTGAGGT
Phenotype
MIM: 115442
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
CSNK2A2 PubMed Links

Gene Details

Gene
CSNK2A2
Gene Name
casein kinase 2 alpha 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001896.2 Intron NP_001887.1
XM_005255800.3 Intron XP_005255857.1
XM_005255801.3 Intron XP_005255858.1
XM_017022945.1 Intron XP_016878434.1

View Full Product Details