Product Details
- SNP ID
-
rs710184
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:37057219 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- AGGATTAAGTGAGATGGTGCCTGGT[A/G]AATGCCTGGTGAGGAGCAGGTGCTC
- Phenotype
-
MIM: 616386
MIM: 609862
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
KCTD17
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2235320] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KCTD17
- Gene Name
- potassium channel tetramerization domain containing 17
- Gene
- TMPRSS6
- Gene Name
- transmembrane protease, serine 6
There are no transcripts associated with this gene.
View Full Product Details