Product Details
- SNP ID
-
rs185926
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:11686437 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TAGGGAAGTATGACAAGAAAAAACA[A/T]CTGTACATATTCAGTACAGACACAA
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF833P
PubMed Links
Gene Details
- Gene
- ZNF833P
- Gene Name
- zinc finger protein 833, pseudogene
There are no transcripts associated with this gene.
View Full Product Details