Product Details

SNP ID
rs12880712
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20991181 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATTTTTTTCTGCAAGTCCACTGGA[G/T]TAGGACCTTCTAAGTTTTTATTCAG
Phenotype
MIM: 616091 MIM: 612166
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC101929718 PubMed Links

Gene Details

Gene
LOC101929718
Gene Name
uncharacterized LOC101929718
There are no transcripts associated with this gene.

Gene
METTL17
Gene Name
methyltransferase like 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029991.1 Intron NP_001025162.1
NM_022734.2 Intron NP_073571.1
XM_006720236.2 Intron XP_006720299.1
XM_017021601.1 Intron XP_016877090.1
Gene
SLC39A2
Gene Name
solute carrier family 39 member 2
There are no transcripts associated with this gene.

View Full Product Details