Product Details

SNP ID
rs9261467
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30136121 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATTGGCATGATATAAGAACCACTC[A/G]TCTGCTGGCCATCTCTCCAGGTCTG
Phenotype
MIM: 616976
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRIM40 PubMed Links

Gene Details

Gene
TRIM40
Gene Name
tripartite motif containing 40
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286633.1 124 UTR 5 NP_001273562.1
NM_138700.4 124 Intron NP_619645.1
XM_011514305.1 124 UTR 5 XP_011512607.1
XM_011514306.1 124 UTR 5 XP_011512608.1
XM_011514308.1 124 UTR 5 XP_011512610.1
XM_011514309.1 124 UTR 5 XP_011512611.1

View Full Product Details