Product Details

SNP ID
rs9261466
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30136063 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGAAAGCATGAGTCTTTCCACCCCC[C/T]GCCCTGAGATTTACTGACATATGAA
Phenotype
MIM: 616976
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
TRIM40 PubMed Links
Additional Information
For this assay, SNP(s) [rs190348808] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
TRIM40
Gene Name
tripartite motif containing 40
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286633.1 66 Intron NP_001273562.1
NM_138700.4 66 Intron NP_619645.1
XM_011514305.1 66 UTR 5 XP_011512607.1
XM_011514306.1 66 UTR 5 XP_011512608.1
XM_011514308.1 66 UTR 5 XP_011512610.1
XM_011514309.1 66 UTR 5 XP_011512611.1

View Full Product Details