Product Details

SNP ID
rs1632937
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:29829776 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTTTAACAGGGTCGGTGGTGAGG[C/G]CTGGGGGTCAGAGACCCTCACCTTC
Phenotype
MIM: 142871
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HLA-G PubMed Links

Gene Details

Gene
HLA-G
Gene Name
major histocompatibility complex, class I, G
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002127.5 Intron NP_002118.1
XM_017010816.1 Intron XP_016866305.1
XM_017010817.1 Intron XP_016866306.1
XM_017010818.1 Intron XP_016866307.1

View Full Product Details