Product Details

SNP ID
rs9663011
Assay Type
Functionally tested
NCBI dbSNP Submissions
44
Location
Chr.1:171143482 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGCACCTCAGTAATCAAATGTTGG[C/G]TCTGATTATAAAGCATTCATGCATT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FMO6P PubMed Links

Gene Details

Gene
FMO6P
Gene Name
flavin containing monooxygenase 6 pseudogene
There are no transcripts associated with this gene.

View Full Product Details