Product Details

SNP ID
rs13218272
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:20102777 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAATTCAAACCCAATACTGTCAT[A/T]TGACACTTAATGACATTTCAGCCAA
Phenotype
MIM: 611732
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MBOAT1 PubMed Links

Gene Details

Gene
MBOAT1
Gene Name
membrane bound O-acyltransferase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080480.2 Intron NP_001073949.1
XM_006714999.2 Intron XP_006715062.1
XM_006715000.3 Intron XP_006715063.1
XM_011514313.2 Intron XP_011512615.1

View Full Product Details