Product Details

SNP ID
rs12191133
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.6:112089133 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
AGCAAGAGAAATGTTTTTGGTTGAA[A/G]TTACTGAAATTTTGGGGTGCAGTTT
Phenotype
MIM: 607345
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM229B PubMed Links

Gene Details

Gene
FAM229B
Gene Name
family with sequence similarity 229 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033564.2 Intron NP_001028736.1
XM_017011174.1 Intron XP_016866663.1
XM_017011175.1 Intron XP_016866664.1
Gene
TUBE1
Gene Name
tubulin epsilon 1
There are no transcripts associated with this gene.

View Full Product Details