Product Details

SNP ID
rs6901629
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:27380029 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCAGAGAACTATAGAATGCTTCCC[C/T]TTCCCCTATACCTTGATACCACATT
Phenotype
MIM: 603282
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF204P PubMed Links

Gene Details

Gene
ZNF204P
Gene Name
zinc finger protein 204, pseudogene
There are no transcripts associated with this gene.

Gene
ZNF391
Gene Name
zinc finger protein 391
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001076781.2 Intron NP_001070249.1
NM_001322288.1 Intron NP_001309217.1
NM_001322289.1 Intron NP_001309218.1
NM_001322293.1 Intron NP_001309222.1
XM_005249082.3 Intron XP_005249139.1
XM_006715086.2 Intron XP_006715149.1
XM_011514569.1 Intron XP_011512871.1

View Full Product Details