Product Details

SNP ID
rs2072875
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.22:50253599 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
CGTGGGGCCCTTCTCTAATCCTAAT[A/G]TGGTCTGAACCACTGAGAGAAGGTT
Phenotype
MIM: 608544 MIM: 602399 MIM: 610053
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HDAC10 PubMed Links

Gene Details

Gene
HDAC10
Gene Name
histone deacetylase 10
There are no transcripts associated with this gene.

Gene
MAPK12
Gene Name
mitogen-activated protein kinase 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303252.1 Intron NP_001290181.1
NM_002969.4 Intron NP_002960.2
Gene
TUBGCP6
Gene Name
tubulin gamma complex associated protein 6
There are no transcripts associated with this gene.

View Full Product Details