Product Details

SNP ID
rs11583313
Assay Type
Functionally tested
NCBI dbSNP Submissions
20
Location
Chr.1:175068559 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGACGACAACTTCTCTCGAAATCC[C/T]CATACTATTACCCTCACAATACCTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TNN PubMed Links

Gene Details

Gene
TNN
Gene Name
tenascin N
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022093.1 Intron NP_071376.1
XM_017002048.1 Intron XP_016857537.1
XM_017002049.1 Intron XP_016857538.1

View Full Product Details