Product Details

SNP ID
rs3734563
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:28381948 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGGTCCCAGAGATGCACCTAAATA[A/G]CAGGAGAATTAAAACCTACTATTTA
Phenotype
MIM: 603978
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZSCAN12 PubMed Links

Gene Details

Gene
ZSCAN12
Gene Name
zinc finger and SCAN domain containing 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163391.1 Intron NP_001156863.1
XM_011515014.2 Intron XP_011513316.1
XM_011515015.2 Intron XP_011513317.1
XM_011515017.2 Intron XP_011513319.1
XM_017011528.1 Intron XP_016867017.1

View Full Product Details