Product Details

SNP ID
rs1048334
Assay Type
Validated
NCBI dbSNP Submissions
64
Location
Chr.1:16014266 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
ATGTGGGAGACGAAACCAAGAGTCA[C/T]TGGGGGCAGCAGGCATTTCCCAGGG
Phenotype
MIM: 602024 MIM: 610692
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C1orf64 PubMed Links

Gene Details

Gene
C1orf64
Gene Name
chromosome 1 open reading frame 64
There are no transcripts associated with this gene.

Gene
CLCNKA
Gene Name
chloride voltage-gated channel Ka
There are no transcripts associated with this gene.

Gene
HSPB7
Gene Name
heat shock protein family B (small) member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014424.4 2654 UTR 3 NP_055239.1
XM_011541249.1 2654 UTR 3 XP_011539551.1
XM_011541250.2 2654 UTR 3 XP_011539552.1

View Full Product Details