Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001301698.1 | Intron | NP_001288627.1 | |||
NM_001301699.1 | Intron | NP_001288628.1 | |||
NM_001301700.1 | Intron | NP_001288629.1 | |||
NM_001301701.1 | Intron | NP_001288630.1 | |||
NM_024066.2 | Intron | NP_076971.1 | |||
XM_005271186.2 | Intron | XP_005271243.1 | |||
XM_006710891.3 | Intron | XP_006710954.1 | |||
XM_017002301.1 | Intron | XP_016857790.1 | |||
XM_017002302.1 | Intron | XP_016857791.1 | |||
XM_017002303.1 | Intron | XP_016857792.1 | |||
XM_017002304.1 | Intron | XP_016857793.1 | |||
XM_017002305.1 | Intron | XP_016857794.1 |