Product Details

SNP ID
rs34011394
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:41308868 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCTGGAACATTATCAGCTTGTTC[C/T]CACCTCAGGGCCATTGCCTGTGCCC
Phenotype
MIM: 602362
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RANGAP1 PubMed Links

Gene Details

Gene
RANGAP1
Gene Name
Ran GTPase activating protein 1
There are no transcripts associated with this gene.

Gene
ZC3H7B
Gene Name
zinc finger CCCH-type containing 7B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017590.5 Intron NP_060060.3
XM_011530016.2 Intron XP_011528318.1

View Full Product Details