Product Details

SNP ID
rs5966747
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:101270472 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTTTCTCTTCCCACCACCCCCTA[A/T]AGATAGGTTTTTCCTTAGGTTCAGA
Phenotype
MIM: 300052 MIM: 300314
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DRP2 PubMed Links

Gene Details

Gene
DRP2
Gene Name
dystrophin related protein 2
There are no transcripts associated with this gene.

Gene
TAF7L
Gene Name
TATA-box binding protein associated factor 7 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168474.1 Intron NP_001161946.1
NM_024885.3 Intron NP_079161.3
XM_005262152.1 Intron XP_005262209.1
XM_006724664.1 Intron XP_006724727.1

View Full Product Details