Product Details

SNP ID
rs734940
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:958953 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGGGGTGTTATGATGGCAGAAGGA[C/T]AGGCAGTGAGGACAGACTTGCTTCC
Phenotype
MIM: 610573
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RSPO4 PubMed Links

Gene Details

Gene
RSPO4
Gene Name
R-spondin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029871.3 2023 UTR 3 NP_001025042.2
NM_001040007.2 2023 UTR 3 NP_001035096.1
XM_017027839.1 2023 Intron XP_016883328.1

View Full Product Details