Product Details

SNP ID
rs8081240
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:28395709 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAAGGGTTTTAGCAGCTCTGTGC[C/T]AGGAACTCTGGGCAAAGGAAAAATA
Phenotype
MIM: 607732 MIM: 611672
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SARM1 PubMed Links

Gene Details

Gene
SARM1
Gene Name
sterile alpha and TIR motif containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015077.3 5425 Intron NP_055892.2
Gene
SLC46A1
Gene Name
solute carrier family 46 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242366.2 5425 UTR 3 NP_001229295.1
NM_080669.5 5425 UTR 3 NP_542400.2
XM_005277786.3 5425 UTR 3 XP_005277843.1
XM_017024110.1 5425 UTR 3 XP_016879599.1

View Full Product Details