Product Details

SNP ID
rs1044732
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.22:19875619 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GGTTTCCACCCAGGTCGTCATCATC[T/C]GGCACCAGGAGCCTGGGGGGGGGTG
Phenotype
MIM: 606448
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
TXNRD2 PubMed Links

Gene Details

Gene
TXNRD2
Gene Name
thioredoxin reductase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282512.1 2018 Intron NP_001269441.1
NM_006440.4 2018 UTR 3 NP_006431.2

View Full Product Details