Product Details

SNP ID
rs4792886
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:45799466 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTTATGACCTCATTTCTTTTTCGC[A/G]TTTAGATTTTCTTCTCTTGCCTTTG
Phenotype
MIM: 122561
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CRHR1 PubMed Links

Gene Details

Gene
CRHR1
Gene Name
corticotropin releasing hormone receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145146.1 Intron NP_001138618.1
NM_001145147.1 Intron NP_001138619.1
NM_001145148.1 Intron NP_001138620.1
NM_001303018.1 Intron NP_001289947.1
NM_001303020.1 Intron NP_001289949.1
NM_004382.4 Intron NP_004373.2
Gene
MGC57346-CRHR1
Gene Name
MGC57346-CRHR1 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256299.2 Intron NP_001243228.1
NM_001303016.1 Intron NP_001289945.1

View Full Product Details