Product Details

SNP ID
rs2189234
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:105154341 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATGTAGTTGAGTTATTTGGTTTAT[G/T]TTATGTCAGAAAATGTCTTACATCT
Phenotype
MIM: 612839
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TET2 PubMed Links

Gene Details

Gene
TET2
Gene Name
tet methylcytosine dioxygenase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127208.2 Intron NP_001120680.1
NM_017628.4 Intron NP_060098.3
XM_005263082.2 Intron XP_005263139.1
XM_006714242.3 Intron XP_006714305.1
XM_017008319.1 Intron XP_016863808.1

View Full Product Details