Product Details

SNP ID
rs11880858
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57584652 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCAATTCTTAGGATGCTAGGAGCC[A/G]TGGAGGGTTGTGTACAGAAGCGGCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZIK1 PubMed Links

Gene Details

Gene
ZIK1
Gene Name
zinc finger protein interacting with K protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010879.3 Intron NP_001010879.2
NM_001321145.1 Intron NP_001308074.1
NM_001321146.1 Intron NP_001308075.1
NM_001321147.1 Intron NP_001308076.1
XM_011526761.1 Intron XP_011525063.1
XM_011526762.1 Intron XP_011525064.1
XM_011526763.1 Intron XP_011525065.1
XM_011526764.1 Intron XP_011525066.1
XM_011526765.2 Intron XP_011525067.1
XM_011526766.2 Intron XP_011525068.1
XM_011526767.2 Intron XP_011525069.1
Gene
ZNF416
Gene Name
zinc finger protein 416
There are no transcripts associated with this gene.

View Full Product Details