Product Details

SNP ID
rs35416930
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:47076016 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGTCCCTGAGAAAAGAGAAGGAG[C/T]TGGAGTGATGATGTGGAACATCTCT
Phenotype
MIM: 615690
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AMIGO2 PubMed Links

Gene Details

Gene
AMIGO2
Gene Name
adhesion molecule with Ig like domain 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143668.1 3540 Intron NP_001137140.1
NM_181847.4 3540 UTR 3 NP_862830.1
XM_005268837.3 3540 Intron XP_005268894.1
Gene
PCED1B
Gene Name
PC-esterase domain containing 1B
There are no transcripts associated with this gene.

View Full Product Details