Product Details

SNP ID
rs1054597
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:1782190 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TTGACTTGCAGCAGAACTGGCTGGA[A/G]CTTCTTCTTCCCAGGAAGGACATCT
Phenotype
MIM: 605866 MIM: 611294
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP8B3 PubMed Links

Gene Details

Gene
ATP8B3
Gene Name
ATPase phospholipid transporting 8B3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178002.2 4797 UTR 3 NP_001171473.1
NM_138813.3 4797 UTR 3 NP_620168.1
XM_006722654.2 4797 UTR 3 XP_006722717.1
XM_006722655.2 4797 UTR 3 XP_006722718.1
XM_006722656.2 4797 UTR 3 XP_006722719.1
XM_011527707.1 4797 UTR 3 XP_011526009.1
XM_011527708.1 4797 UTR 3 XP_011526010.1
XM_011527709.1 4797 UTR 3 XP_011526011.1
XM_011527710.2 4797 UTR 3 XP_011526012.1
XM_011527711.2 4797 Intron XP_011526013.1
XM_011527712.1 4797 UTR 3 XP_011526014.1
Gene
ONECUT3
Gene Name
one cut homeobox 3
There are no transcripts associated with this gene.

View Full Product Details