Product Details
- SNP ID
-
rs2107654
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:65636955 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGATAGTACAGACTAAGCACTGGGA[C/T]GAGAAGGGGGGTGACTTCTTAAAGA
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CEP112
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs142554648] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CEP112
- Gene Name
- centrosomal protein 112
View Full Product Details