Product Details

SNP ID
rs11982634
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:39626668 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGGGCATTTTTCCCGTGGGTGTCC[C/T]GGTAAAGTATTTGGACTCAATTCTG
Phenotype
MIM: 179550
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RALA PubMed Links

Gene Details

Gene
RALA
Gene Name
RALA Ras like proto-oncogene A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005402.3 Intron NP_005393.2
XM_006715762.2 Intron XP_006715825.1
XM_011515466.1 Intron XP_011513768.1
XM_011515467.1 Intron XP_011513769.1

View Full Product Details