Product Details

SNP ID
rs2075762
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:18879050 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AAACGCCACGATGTACTGCGAGAGG[A/G]GAGGGGAGGTGCCAGTGAGAAGAAA
Phenotype
MIM: 606919 MIM: 602880
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CERS1 PubMed Links

Gene Details

Gene
CERS1
Gene Name
ceramide synthase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290265.1 Intron NP_001277194.1
NM_021267.4 Intron NP_067090.1
NM_198207.2 Intron NP_937850.1
Gene
GDF1
Gene Name
growth differentiation factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001492.5 Intron NP_001483.3

View Full Product Details