Product Details

SNP ID
rs34955667
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:39680834 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAAGGTGTCCCCTTCTGGAAAGGA[C/T]GTCCATGGCCCTTGATGATTGTGAT
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
LGALS17A PubMed Links
Additional Information
For this assay, SNP(s) [rs74921025] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
LGALS17A
Gene Name
galectin 14 pseudogene
There are no transcripts associated with this gene.

View Full Product Details