Product Details

SNP ID
rs16973566
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:39686277 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGAATGACAGTCCTTTTCCCTCTA[C/T]AAGAGCTGAGATCACCTGCTTCATA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LGALS17A PubMed Links

Gene Details

Gene
LGALS17A
Gene Name
galectin 14 pseudogene
There are no transcripts associated with this gene.

View Full Product Details