Product Details

SNP ID
rs2240633
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:43624285 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CACAGGACCAGGTCATCAGAGGACG[A/G]TCCCAGTCTGGCCCCATCGCTGCCA
Phenotype
MIM: 609744
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CADM4 PubMed Links

Gene Details

Gene
CADM4
Gene Name
cell adhesion molecule 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145296.1 Intron NP_660339.1
XM_005258620.2 Intron XP_005258677.1
XM_017026452.1 Intron XP_016881941.1
Gene
SRRM5
Gene Name
serine/arginine repetitive matrix 5
There are no transcripts associated with this gene.

Gene
ZNF428
Gene Name
zinc finger protein 428
There are no transcripts associated with this gene.

View Full Product Details