Product Details

SNP ID
rs4802866
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51796422 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTAACTTAGAAGTCTGGAAGCCATC[A/G]GAAGGTATTGCGAATAACATCACCA
Phenotype
MIM: 136539
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FPR3 PubMed Links

Gene Details

Gene
FPR3
Gene Name
formyl peptide receptor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002030.4 Intron NP_002021.3
XM_011526687.2 Intron XP_011524989.1

View Full Product Details