Product Details

SNP ID
rs712696
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:127347041 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGCATGGCTTCAGTGCAAACCCAG[C/T]AGTGTATGCCTTGGGAAGGCAGGGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF800 PubMed Links

Gene Details

Gene
ZNF800
Gene Name
zinc finger protein 800
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_176814.4 3799 Intron NP_789784.2
XM_005250181.3 3799 Intron XP_005250238.1
XM_006715867.3 3799 UTR 3 XP_006715930.1
XM_006715868.3 3799 UTR 3 XP_006715931.1
XM_006715869.3 3799 UTR 3 XP_006715932.1
XM_011515859.2 3799 UTR 3 XP_011514161.1

View Full Product Details