Product Details

SNP ID
rs33964243
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35903370 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACAGTGGCTGCAGCTCAGACGAC[C/T]CCAGGAGAGAGATCATCACTCCCTG
Phenotype
MIM: 604089 MIM: 604142
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HCST PubMed Links

Gene Details

Gene
HCST
Gene Name
hematopoietic cell signal transducer
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007469.1 177 Silent Mutation ACC,ACT T21T NP_001007470.1
NM_014266.3 177 Silent Mutation ACC,ACT T21T NP_055081.1
XM_017026193.1 177 Silent Mutation ACC,ACT T21T XP_016881682.1
Gene
NFKBID
Gene Name
NFKB inhibitor delta
There are no transcripts associated with this gene.

Gene
TYROBP
Gene Name
TYRO protein tyrosine kinase binding protein
There are no transcripts associated with this gene.

View Full Product Details