Product Details

SNP ID
rs732479
Assay Type
Validated
NCBI dbSNP Submissions
29
Location
Chr.1:226877777 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
ACACTTGTTTTCTGGTGAGGAGGCC[G/A]CATGTATGAATGACGTTTGTGGGTT
Phenotype
MIM: 600759
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
PSEN2 PubMed Links

Gene Details

Gene
PSEN2
Gene Name
presenilin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000447.2 Intron NP_000438.2
NM_012486.2 Intron NP_036618.2
XM_005273199.3 Intron XP_005273256.1
XM_017001835.1 Intron XP_016857324.1
XM_017001836.1 Intron XP_016857325.1

View Full Product Details