Product Details
- SNP ID
-
rs4780822
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:19716676 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACTTCAAGAAATACAGTAGCTCGCC[A/G]CCTAATCCCAGGTTTCCGTTACCCC
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
IQCK
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112819134] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- IQCK
- Gene Name
- IQ motif containing K
- Gene
- KNOP1
- Gene Name
- lysine rich nucleolar protein 1
View Full Product Details