Product Details

SNP ID
rs34777281
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:44624382 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACTTGTGCTGCTAAGTTGAGGCC[A/C]TTGACGGCTTCCCAGACTGTTAAGA
Phenotype
MIM: 613022
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OGDH PubMed Links

Gene Details

Gene
OGDH
Gene Name
oxoglutarate dehydrogenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003941.2 130 Silent Mutation CCA,CCC P13P NP_001003941.1
NM_001165036.1 130 Silent Mutation CCA,CCC P13P NP_001158508.1
NM_002541.3 130 Silent Mutation CCA,CCC P13P NP_002532.2
XM_005249759.4 130 Silent Mutation CCA,CCC P13P XP_005249816.1
XM_005249761.3 130 Silent Mutation CCA,CCC P13P XP_005249818.1
XM_011515408.2 130 Silent Mutation CCA,CCC P13P XP_011513710.1

View Full Product Details