Product Details

SNP ID
rs13240413
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:101128820 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCACTACACTCCAAGTCACTGGA[C/T]CCTTGAATCTCCAAGGGTGTCTGAC
Phenotype
MIM: 173360
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SERPINE1 PubMed Links

Gene Details

Gene
SERPINE1
Gene Name
serpin family E member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000602.4 Intron NP_000593.1
XM_017012260.1 Intron XP_016867749.1

View Full Product Details