Product Details

SNP ID
rs2270073
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.14:73852051 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CAGGGTGACTTCTAGGCGTCCTCCC[G/T]GAGCTACCGGGGACACTGGCGTTAT
Phenotype
MIM: 608642
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PTGR2 PubMed Links

Gene Details

Gene
PTGR2
Gene Name
prostaglandin reductase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146154.1 134 Intron NP_001139626.1
NM_001146155.1 134 Intron NP_001139627.1
NM_152444.2 134 UTR 5 NP_689657.1

View Full Product Details