Product Details
- SNP ID
-
rs2079431
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:31168348 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TATTTGTCTTCCTTTGACTGACTTA[C/T]TTCACCTAACATGATGTTCTTGAGG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MIR3928
PubMed Links
Gene Details
- Gene
- MIR3928
- Gene Name
- microRNA 3928
There are no transcripts associated with this gene.
- Gene
- RNF185
- Gene Name
- ring finger protein 185
View Full Product Details