Product Details

SNP ID
rs35480486
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:70662479 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGGAAGGTCAGGGGAAACCTGAC[C/T]ACTGGACAGGAGTGAGGCAGGTACA
Phenotype
MIM: 102681
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADD2 PubMed Links

Gene Details

Gene
ADD2
Gene Name
adducin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185054.1 3392 UTR 3 NP_001171983.1
NM_001185055.1 3392 Intron NP_001171984.1
NM_001617.3 3392 UTR 3 NP_001608.1
NM_017482.3 3392 Intron NP_059516.2
NM_017488.3 3392 UTR 3 NP_059522.1
XM_011532502.2 3392 UTR 3 XP_011530804.1

View Full Product Details