Product Details

SNP ID
rs34676576
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:74499675 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGCAGAGCTGGCCAGGGAGTAGG[C/T]ACAGCCTCATCTCTAATATTTAGAC
Phenotype
MIM: 615317 MIM: 602091 MIM: 601015
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ISCA2 PubMed Links

Gene Details

Gene
ISCA2
Gene Name
iron-sulfur cluster assembly 2
There are no transcripts associated with this gene.

Gene
LTBP2
Gene Name
latent transforming growth factor beta binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000428.2 7062 UTR 3 NP_000419.1
XM_011536765.1 7062 UTR 3 XP_011535067.1
Gene
NPC2
Gene Name
NPC intracellular cholesterol transporter 2
There are no transcripts associated with this gene.

View Full Product Details