Product Details

SNP ID
rs11615364
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:4651496 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCACAGTTTCTAGAATAATTCTTG[G/T]CCCATAATTATTAAAAAAAAACAAT
Phenotype
MIM: 604689 MIM: 603834
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
AKAP3 PubMed Links

Gene Details

Gene
AKAP3
Gene Name
A-kinase anchoring protein 3
There are no transcripts associated with this gene.

Gene
NDUFA9
Gene Name
NADH:ubiquinone oxidoreductase subunit A9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005002.4 Intron NP_004993.1

View Full Product Details