Product Details

SNP ID
rs2505327
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.10:27399036 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ATAAAAATAGCAAAATAACAGGGTC[A/G]TTCCTGTATAGATGCAAAAACATTG
Phenotype
MIM: 611791
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PTCHD3 PubMed Links

Gene Details

Gene
PTCHD3
Gene Name
patched domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001034842.3 1680 Missense Mutation ACG,ATG T521M NP_001030014.2

View Full Product Details