Product Details

SNP ID
rs3785832
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:61461433 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGCTGGGCCCTGGCCTCTTCCCCG[C/T]ACACCCAGGATTCCTCCTGAAAAGC
Phenotype
MIM: 601719
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TBX4 PubMed Links

Gene Details

Gene
TBX4
Gene Name
T-box 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321120.1 Intron NP_001308049.1
NM_018488.3 Intron NP_060958.2
XM_011525490.2 Intron XP_011523792.1
XM_011525491.2 Intron XP_011523793.1
XM_011525495.2 Intron XP_011523797.1

View Full Product Details