Product Details

SNP ID
rs11731468
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.4:13295056 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACAAGTCAGGGGAGGCAGGGAGTG[C/T]GAACACAGTACATGGGAGGGGGAGG
Phenotype
MIM: 612994
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
RAB28 PubMed Links

Gene Details

Gene
RAB28
Gene Name
RAB28, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017979.2 1776 Intron NP_001017979.1
NM_001159601.1 1776 Intron NP_001153073.1
NM_004249.3 1776 Intron NP_004240.2
XM_005248218.4 1776 Intron XP_005248275.1
XM_011513911.2 1776 UTR 3 XP_011512213.1
XM_011513912.2 1776 Intron XP_011512214.1
XM_017008850.1 1776 Intron XP_016864339.1

View Full Product Details