Product Details

SNP ID
rs2191915
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:209777963 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCTGATTGCTCTATTCTCCTTTCC[C/T]CGCTCTATTCCTGTCATAGTCCTCA
Phenotype
MIM: 612636
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UNC80 PubMed Links

Gene Details

Gene
UNC80
Gene Name
unc-80 homolog, NALCN activator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032504.1 Intron NP_115893.1
NM_182587.3 Intron NP_872393.3
XM_005246476.1 Intron XP_005246533.1
XM_011511004.1 Intron XP_011509306.1
XM_011511005.1 Intron XP_011509307.1
XM_011511006.1 Intron XP_011509308.1
XM_011511007.1 Intron XP_011509309.1
XM_011511008.1 Intron XP_011509310.1
XM_011511010.2 Intron XP_011509312.1
XM_011511012.1 Intron XP_011509314.1
XM_017003884.1 Intron XP_016859373.1
XM_017003885.1 Intron XP_016859374.1
XM_017003886.1 Intron XP_016859375.1
XM_017003887.1 Intron XP_016859376.1
XM_017003888.1 Intron XP_016859377.1
XM_017003889.1 Intron XP_016859378.1
XM_017003890.1 Intron XP_016859379.1
XM_017003891.1 Intron XP_016859380.1
XM_017003892.1 Intron XP_016859381.1
XM_017003893.1 Intron XP_016859382.1
XM_017003894.1 Intron XP_016859383.1

View Full Product Details