Product Details

SNP ID
rs2302626
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:30022586 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACTTCCAGGGGAAAAAAAAAAGTT[A/G]TAGTGACTCTAACTTCTGTCTCCTA
Phenotype
MIM: 614505 MIM: 608639
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FKBP14 PubMed Links

Gene Details

Gene
FKBP14
Gene Name
FK506 binding protein 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017946.3 Intron NP_060416.1
Gene
PLEKHA8
Gene Name
pleckstrin homology domain containing A8
There are no transcripts associated with this gene.

View Full Product Details