Product Details

SNP ID
rs12314703
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:14839292 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTCCCATATCCTACACATCCTACA[C/T]ACCAGCCATCCAGAATTTCAGAGTC
Phenotype
MIM: 110600
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ART4 PubMed Links

Gene Details

Gene
ART4
Gene Name
ADP-ribosyltransferase 4 (Dombrock blood group)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021071.2 Intron NP_066549.2

View Full Product Details